Learn More
SC144 hydrochloride, MedChemExpress
Product Code. 16476836
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16476836
Brand: Medchem Express HY15614A
MedChemExpress SC144 hydrochloride is a first-in-class, orally active gp130 (IL6-beta) inhibitor.
SC144 hydrochloride binds gp130, induces gp130 phosphorylation (S782) and deglycosylation, abrogates Stat3 phosphorylation and nuclear translocation, and further inhibits the expression of downstream target genes. SC144 hydrochloride shows potent inhibition of gp130 ligand-triggered signaling. SC144 hydrochloride induces apoptosis in human ovarian cancer cells.
Specifications
Chemical Name or Material SC144 hydrochloride
CAS 917497-70-2
Color Gray
Purity Grade Notes Research
Molecular Formula C16H12ClFN6O
Quantity 100 mg
Solubility Information DMSO : 10 mg/mL (27.87 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C1=NC=CN=C1)NNC2=NC3=C(N4C2=CC=C4)C=CC(F)=C3.[H]Cl
Molecular Weight (g/mol) 358.76
Formula Weight 358.76
Percent Purity 98.89%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.