missing translation for 'onlineSavingsMsg'
Learn More

SB-616234-A, MedChemExpress

Product Code. p-200100486
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16480238

Brand: Medchem Express HY19477

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, with anxiolytic and antidepressant activity.

SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, with anxiolytic and antidepressant activity.

Specifications

Chemical Name or Material SB-616234-A
CAS 908601-49-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C32H36ClN5O3
Quantity 5 mg
Solubility Information DMSO : 10 mg/mL (17.42 mM; Need ultrasonic)
SMILES O=C(N1CCC2=C1C=C(N3C[C@@H](C)N[C@@H](C)C3)C(OC)=C2)C4=CC=C(C5=CC=C(C6=NOC(C)=N6)C=C5C)C=C4.[H]Cl
Molecular Weight (g/mol) 574.11
Formula Weight 574.11
Percent Purity 98.14%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.