Learn More
SB-616234-A, MedChemExpress
Product Code. 16480238
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16480238
Brand: Medchem Express HY19477
MedChemExpress SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, with anxiolytic and antidepressant activity.
SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, with anxiolytic and antidepressant activity.
Specifications
Chemical Name or Material SB-616234-A
CAS 908601-49-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C32H36ClN5O3
Quantity 5 mg
Solubility Information DMSO : 10 mg/mL (17.42 mM; Need ultrasonic)
SMILES O=C(N1CCC2=C1C=C(N3C[C@@H](C)N[C@@H](C)C3)C(OC)=C2)C4=CC=C(C5=CC=C(C6=NOC(C)=N6)C=C5C)C=C4.[H]Cl
Molecular Weight (g/mol) 574.11
Formula Weight 574.11
Percent Purity 98.14%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.