Learn More
SB-366791, MedChemExpress
Product Code. 16471277
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16471277
Brand: Medchem Express HY12245
MedChemExpress SB-366791 is a potent and selective vanilloid receptor (VR1/TRPV1) antagonist (IC50=5.7 nM). SB-366791 can be used for the research of inflammation.
SB-366791 is a potent and selective vanilloid receptor (VR1/TRPV1) antagonist (IC50=5.7 nM). SB-366791 can be used for the research of inflammation.
Specifications
Chemical Name or Material SB-366791
CAS 472981-92-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H14ClNO2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (347.54 mM)
SMILES O=C(NC1=CC=CC(OC)=C1)/C=C/C2=CC=C(Cl)C=C2
Molecular Weight (g/mol) 287.74
Formula Weight 287.74
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.