Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Shop All Thermo Scientific ProductsDescription
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Specifications
| Description | Centrifuge Tubes: 6 x 50mL conical, Medium capacity models |
| Type | Rotor |
| For Use With (Equipment) | SPD1030, SPD120, SPD130, SPD140DDA |
| Holds | 6 x 50 mL Conical Tubes |
| Model | RH6-50 |
| Rotor System | Traditional |
| Volume (Metric) | 50 mL |