Learn More
Berkshire International SatPax™ 670 Pre-wetted 55% Cellulose / 45% Polyester Non woven Cleanroom Wiper
Product Code. 18888956 Shop All Berkshire Products
Change view
Click to view available options
Length (English):
11 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English)
18888956 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18888956 Supplier Berkshire International Supplier No. SPX67091112BPR
  • ONLINE EXCLUSIVE PRICE
    2840.01 NOK
    1915.52 NOK / Pack of 12
    Save 924.49 NOK
    32% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
Pre-wetted nonwoven cleanroom wipers made with a hydroentangled cellulose and polyester blend for cleanroom wiping.
  • Low extractables and fibre and particle counts
  • Smooth and durable with good wet strength
  • Reduces alcohol usage and preparation / handling costs
  • Reduces VOC emissions
  • Increases cleaning efficiency
  • Increases cleaning protocol consistency
  • Lot traceability
Specifications
Product Type Cleanroom Wiper
Length (English) 11 in.
Length (Metric) 28 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Polyester Cellulose
Autoclavable Not Autoclavable
Disposable Single-use
Ply 1-ply
Sterility Non-sterile
compliance-icons compliance-icons
Product Identifier
  • Satpax 670 Cleanroom Presaturated Wipes, 25 per pack
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: Isopropanol