Learn More
Sartorius Tygon Tubing for Vivaflow™ 50
Tubing Set for Vivaflow™ 50 Comprises: 2 x 1m size 16 PVC tubing with inlet fittings, 2 x 50 cm size 16 PVC tubing with 0.6 mm flow restrictors, 1 x series interconnector
Brand: Sartorius VFA034
1596.45 NOK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Size: 16 PVC tubing with 0.6 mm flow restrictor; 1x series interconnector.Specifications
VF50 Tubing Kit | |
Vivaflow™ 50 | |
2 x 1m Size 16 PVC tubing with inlet fittings, 2 x 50cm size 16 PVC tubing with 0.6mm flow restrictors, 1x series interconnector |