Includes
Handbook, aluminum case, stainless-steel tweezers, filtration system for samples, filtration systems for sterile water, inoculation loop, culture media (nutrient broth), wort nutrient pad sets, standard nutrient pad sets, Endo nutrient pad sets. Filtration system includes device, 3-way valve, adapter, syringe and glass fiber filter. Filtration system includes filter holder, syring, and membrane filter.
All the equipment necessary for microbiological testing contained in a rugged aluminum case.
- Seven easy-to-understand experiments
- Compact and robust design
- Portable for added convenience
The handbook included in the case provides general instructions and detailed descriptions of methods for the experiments.
Topics:
- Detection of microorganisms in water, air and soil
- Effects of antibiotics
- Detection of yeasts on substrates in nature
- Production of gas through alcoholic fermentation
- Bacterial growth at different temperatures
Specifications
Specifications
| Product Type | School Microbiology Kit |
| For Use With (Application) | Ideal teaching aid for instruction in microbiology and environmental protection in schools and other educational institutions |
| Includes | Handbook, aluminum case, Stainless Steel tweezers, filtration system for samples, filtration systems for sterile water, inoculation loop, culture media (nutrient broth), wort nutrient pad sets, standard nutrient pad sets, endo nutrient pad sets, filtration system includes device, 3 way valve, adapter, syringe and Glass fiber filter, filtration system includes filter holder, syringe, and membrane filter |