Learn More
Sartorius Pressure Heads for Vivaspin™ 20 and Vivacell™ 70, 100, 250
Shop All Sartorius Products
Click to view available options
For Use With:
Vivacell 70 pressure fuge
Vivacell™ 100
Vivacell™ 250
Vivacell™ 70
Vivaspin™ 20
Unit Size:
Each
Description
The Vivaspin™ 20 is an ultrafiltration unit with unique twin membrane for sample concentration with high process flexibility: centrifugation or centrifugation of pressurized units, as well as pressure filtration in stationary benchtop mode.
Specifications
Specifications
| For Use With | Vivaspin™ 20 |
| Type | Pressure Head |