Learn More
Sartorius Pressure Heads for Vivaspin™ 20 and Vivacell™ 70, 100, 250
Product Code. 10537441 Shop All Sartorius Products
Change view
Click to view available options
For Use With:
Vivacell 70 pressure fuge
Vivacell™ 100
Vivacell™ 250
Vivacell™ 70
Vivaspin™ 20
Unit Size:
Each
Product Code. For Use With unitSize
10537441 Vivacell™ 70 Each
11705036 Vivacell™ 250 Each
11790956 Vivaspin™ 20 Each
11735036 Vivacell 70 pressure fuge Each
10609731 Vivacell™ 100 Each
5 options
Product Code. 10537441
Brand: Sartorius VCA700
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For various Vivacell models
Vivacell™ 70 is a disposable ultrafiltration unit for the concentration of samples between 10 ml and 70 ml and can be used either in a centrifuge or for pressure filtration. Vivacell™ 70 thus combines the ease of use of centrifugal devices with the flexibility and control provided by pressurized ultrafiltration cells. Vivacell™ 70 is easy to assemble, requires no tubing connections or stirring mechanisms and can be adapted to equipment availability or to specific user preferences. For convenience, the units are simply spun in a large capacity centrifuge (carriers accepting 250ml bottles).
For even faster concentration, particularly with difficult samples, the device can be pressurized with air or inert gas before centrifuging. For better process control or for handling of single samples, gas pressure can be combined with gentle orbital shaking. For highest simplicity of use with minimum equipment requirements, the device is pressurized and then left standing on a bench top or in a refrigerator. The concentrator features a vertical membrane that minimizes fouling, while the built-in deadstop of 150 μL will prevent concentration to dryness. Easy to handle. High flux PES membrane Vertical membrane design for ultra-fast concentration. High retentate recovery >90%. Total process flexibility. Universal rotor compatibility.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Vivacell™ 70
Includes Reservoir and filtrate bottle
Type Pressure Head