Description
Special feed roll technology makes each filter quickly and reliably accessible; avoids filter band slippage or even damaged membranes.
- Compact and robust design
- Works hands-free by an optical sensor (when sensor detects approaching tweezers) or touch button
- Rapid and reliable transport due to sprocket feed roll technology
- Controller prevents unwanted dispensing of several membrane filters at a time
- Easy insertion of the filter band
- Clear, compact design of the dispenser allows quick and easy cleaning
- Low weight for easy transport
- Interface port available so that other sensor systems can be connected to control the dispenser
Order Info
Accessory pedal switch available: 1ZE---0028. Membrane filters available separately.
Specifications
Specifications
| Certifications/Compliance | CE and EMC, EN 50081-1 and -2, EN 50082-1 and -2, EN 61010 |
| For Use With (Equipment) | Membrane Filter |
| For Use With (Application) | To prevent unwanted dispensing of several membrane filters |
| Type | Membrane Filter Dispenser |
| Width (English) | 6.5 in. |
| Width (Metric) | 16.5 cm |
| Weight (Metric) | 2.9 kg |