Description
- Vaccuum Filter Holder is delivered with a glass frit filter support that ensures a uniform distribution of the retained particles; therefore it is recommended when the residue on the filter surface is of interest.
- Millex Filter is delivered with a PTFE-coated screen filter support that has the benefit to be easy to clean. Therefore it is recommended when the filtrate is required or when liquids that are difficult to remove from glass supports must be examined.
Specifications
Specifications
| Disposable | Reusable |
| For Use With (Equipment) | Vacuum Filter Holder, Manifold System |
| Material | Glass |
| Sterility | Sterile |
| Filtration Area | 3 cm2 |
| Quantity | Each |
| Type | Millex Samplicity Filter |
| Volume (Metric) | 30 mL |
| Diameter (Metric) Membrane | 2.5 cm |