Learn More
Sartorius Entris™ Precision Balances
Shop All Sartorius Products
Click to view available options
Capacity:
150 g
320 g
420 g
620 g
Model:
ENTRIS153-1S
ENTRIS153i-1S
ENTRIS323-1S
ENTRIS323i-1S
ENTRIS423-1S
ENTRIS423i-1S
ENTRIS623-1S
ENTRIS623i-1S
Stabilization Time:
1.0 sec.
1.1 sec.
1.3 sec.
Unit Size:
Each
Beschreibung
- White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
- New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
- RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
- Total ease of operation
- All keys have positive click action
- Clearly defined control panel
- Monolithic weighing cell provides high thermal stability and extremely fast response times
- Fast response time and display stability
- Language-driven user interface
- Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Spezifikation
Spezifikation
| Calibration | Internal |
| Capacity | 320 g |
| Readability | 1 mg |
| Linearity | 2 mg |
| Model | ENTRIS323i-1S |
| Repeatability | 1 mg |
| Stabilization Time | 1.1 sec. |
| Type | Precision Balance |
| Dimensions (D x W x H) | 230 x 303 x 136 mm |
| Diameter (Metric) Outer Pan | 115 mm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur