Learn More
Sartorius Interface Module for Cubis Balances
GREENER_CHOICE
Product Code. p-4235234 Shop All Sartorius Products
Click to view available options
Connection Type:
Option P: RS-232 9 Pin, PS/2
For Use With (Application):
Connecting a PC or a keyboard
Unit Size:
Each
Product Code. 15385601
Brand: Sartorius YDO01MSP
Interface Module for use with Cubis balances.
  • Option R (YDO01MS-R): RS-232C data interface, 25-pin, for connection of Cubis accessory equipment
  • Option B (YDO01MS-B): Bluetooth™ interface module for wireless connection of the YDP10BT data printer
  • Option P (YDO01MS-P): RS-232C data interface, 9-pin, including PS/2 port for connecting a PC or a keyboard
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cubis Balance
Connection Type Option P: RS-232 9 Pin, PS/2
For Use With (Application) Connecting a PC or a keyboard
Type Data Interface