Learn More
Sartorius Combisart™ Accessories for Colony Counting
Shop All Sartorius Products
Click to view available options
Outlet Style:
Hose Nipple, DN 10
TR 20 x 2mm male thread with DN 24 (≈24mm) hexagonal nut
Unit Size:
Each
Description
For use with Combisart Individual and Multibranch Systems for Colony Counting (Rep. Cat. Nos. 14-555-800 and 14-555-802).
Specifications
Specifications
| Product Type | Base Support |
| Filtration Area | 12.5cm2 |
| Inlet Style | Female Threads, TR 20 x 2 |
| Material | Stainless Steel |
| Outlet Style | Hose Nipple, DN 10 |
| Sample Size | 3 or 6 |
| Certifications/Compliance | ISO 8199, B.S. 304S31, AISI 304 |
| For Use With (Application) | Microbiological analysis or particle count in quality assurance |
| Includes | 50mm Frit |