Learn More
Sartorius Balance Data Transfer Cables
GREENER_CHOICE
Product Code. 10647893 Shop All Sartorius Products
Change view
Click to view available options
Connection Type:
RS-232 25 Pin to USB
Type:
RS232 to USB Cable
Length (English):
6.6 ft.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Type Length (English) Unit Size
10647893 RS-232 25 Pin to USB RS232 to USB Cable 6.6 ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10647893 Supplier Sartorius Supplier No. YCC01USBM2
In Stock
Add to basket
Add to basket
RS-232 Balance Data Transfer Cables in a variety of lengths.
RS-232 cables in a variety of lengths
  • For use with Sartorius balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CPA Balance, Extended Balance, Master Pro LA Balance
Connection Type RS-232 25 Pin to USB
Length (English) 6.6 ft.
Length (Metric) 2 m
Type RS232 to USB Cable