Learn More
Sartorius Accessories for Combisart™ Systems: Prefilter Attachment
Prefilter attachment for use with Sartorius Combisart Systems
Brand: Sartorius 16807
Additional Details : Weight : 0.31500kg
Description
- Stainless steel
- Holds prefilter membrane for bacteriological analyses
Specifications
Prefilter Attachment | |
Stainless Steel | |
Microbiological analysis | |
Combisart™ system |