Learn More
Sartorius Accessories for Combisart™ Systems: Prefilter Attachment
Prefilter attachment for use with Sartorius Combisart Systems
Brand:  Sartorius 16807
Additional Details : Weight : 0.31500kg
Product Code. 15982540
  • 4040.00 NOK / Each
Estimated Shipment: 10-07-2024
Description
  • Stainless steel
  • Holds prefilter membrane for bacteriological analyses
Specifications
Specifications
Prefilter Attachment
Stainless Steel
Microbiological analysis
Combisart™ system