Learn More
Sartorius Qualitative Grade 39/N Filter Papers
This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.
Brand:  Sartorius FT-3-483-385
Additional Details : Weight : 1.04800kg
 View more versions of this product
Product Code. 15992950
  • 2420.00 NOK / Pack of 100
Estimated Shipment: 10-06-2024
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.
Specifications
Specifications
Disc
39/N
0.65 mm
5 sec.
50/Pk.
385 mm