Learn More
Sartorius Qualitative Grade 1290 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand:  Sartorius FT-3-208-110
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10032361
  • 186.00 NOK / Pack of 100
Estimated Shipment: 19-06-2024
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
100 sec.
100/Pk.
110 mm
3 to 5 μm
1290
0.21 mm