Learn More
Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle
BIOPHARMAGREENER_CHOICE
Product Code. p-7237799 Shop All Sartorius Products
Change view
Click to view available options
Sample Volume:
150 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Product Code. Sample Volume unitSize
17404412 150 mL Pack of 12
17414412 250 mL Pack of 12
17424412 500 mL Pack of 12
17434412 1000 mL Pack of 12
4 options
Product Code. 17404412
Brand: Sartorius 18022E
Only null left
Add to basket
Add to basket
Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.
  • The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
For Use With (Equipment) Sartolab
Material Polystyrene
Material (Membrane) Polystyrene
Sample Volume 150 mL