Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB105801R-100
 View more versions of this product
Product Code. 18772527
  • 5135.00 NOK / 100µg
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Blocking Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spike RBD
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Mouse
Protein A or G purified from cell culture supernatant
RUO
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a
Blocking Assay
1035419R
Blockade of Receptor-ligand Interaction
YP_009724390.1
Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD, Ala319-Phe541, Accession # YP_009724390.1
100 μg
Primary
SARS-CoV-2