missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein RBD Omicron Recombinant Rabbit Monoclonal Antibody (HL1866)
GREENER_CHOICE

Product Code. 17829245
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17829245 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17829245 Supplier Invitrogen™ Supplier No. MA547207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) variant Omicron (B 1 1 529) has attracted great concerns since its identification in South Africa Omicron is the fifth variant of concern (VOC) after Alpha (B 1 1 7), Beta (B 1 351), Gamma (P 1) and Delta (B 1 617 2) Omicron variant harbors >30 substitutions/deletions/insertions in the spike, including 15 substitutions in receptor-binding domain (RBD) [PMID: 35397623].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike Protein RBD Omicron
Applications ELISA, Inhibition Assays, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1866
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias 2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike
Host Species Rabbit
Immunogen Full length SARS-coV-2 RBD recombinant protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.