missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Protein Monoclonal Antibody (OTI5A7), TrueMAB™, OriGene
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 Spike Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein in Virus samples. It is validated for ELISA
Specifications
Specifications
| Antigen | SARS-CoV-2 Spike Protein |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | OTI5A7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene Alias | Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 |
| Host Species | Mouse |
| Immunogen | Recombinant fusion protein of SARS-CoV-2 Spike RBD protein and virion surface domain of membrane protein (M-protein) produced in HEK293 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?