Learn More
ACROBiosystems SARS-CoV-2 S1 protein (HV69-70del, Y144del, N501Y, A570D, D614G, P681H), His Tag
Brand:  ACROBiosystems S1N-C52Hr-100ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17206480
  • 4442.46 NOK / 100µg
Estimated Shipment: 16-06-2023
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.5 kDa.
Specifications
Specifications
Powder
100 μg
Val 16 - Arg 685
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Unconjugated
HEK293
SARS-CoV-2
76.5 kDa
SARS-CoV-2 S1 protein, His Tag (S1N-C52Hr) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1).
-20°C
S1 protein
His Tag
Viral Proteins