missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems SARS-CoV-2 S protein (D614G), His Tag, Super stable trimer

MALS verified

Brand:  ACROBiosystems SPN-C52H3-50ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17106771

  • 5239.82 NOK / 50µg
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 138.0 kDa. The protein migrates as 170-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

QHD43416.1 (D614G)
50 µg
-20°C
S protein
SARS-CoV-2
PBS
HEK293
Viral Proteins
138.0 kDa
SARS-CoV-2 S protein (D614G), His Tag, Super stable trimer (SPN-C52H3) is the ectodomain of SARS-CoV-2 S protein that contains AA Val 16 - Pro 1213 (Accession # QHD43416.1) and D614G mutation, which has become increasingly common in SARS-CoV-2 viruses from around the world . The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively.)
Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19
Unconjugated
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers