missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARDH Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22762-1-AP-20UL
This item is not returnable.
View return policy
Description
The function remains known.Specifications
| SARDH | |
| Polyclonal | |
| Unconjugated | |
| SARDH | |
| BPR 2, DMGDHL1, SAR, sarcosine dehydrogenase, SARD, SARDH, SDH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114123, 1757 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q64380, Q9UL12 | |
| SARDH | |
| SARDH Fusion Protein Ag18738 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction