missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine oxidase Antibody, Novus Biologicals™

Product Code. 18221033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18221033 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18221033 Supplier Novus Biologicals Supplier No. NB600649100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Sarcosine oxidase Polyclonal specifically detects Sarcosine oxidase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pipecolic acid oxidase
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias EC 1.5.3.1, EC 1.5.3.7, L-pipecolate oxidase, L-pipecolic acid oxidase, LPIPOXperoxisomal sarcosine oxidase, pipecolic acid oxidase, PSO
Gene Symbols PIPOX
Host Species Goat
Immunogen Sarcosine Oxidase (Microbial)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cell Biology, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51268
Test Specificity Assay by immunoelectrophoresis resulted in a single precipition arc against anti-Goat Serum as well as purified and partially purified Sarcosine Oxidase (Microbial).
Target Species Bacteria
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.