missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine ethyl ester hydrochloride, 98+%

Product Code. 11464454
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11464454

Brand: Thermo Scientific Alfa Aesar A13112.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52605-49-9 | C5H12ClNO2 | 153.61 g/mol

Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sarcosine ethyl ester hydrochloride, It is used as intermediate in the synthesis of antienzyme agents for toothpaste. Found to be a marker for prostate cancer bioagression. It can be used to produce N-methyl-N-nitroso-glycine ethyl ester. This reaction will need reagent NaNO2, and the menstruum H2O and aq. acetic acid.

Solubility
Soluble in water 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52605-49-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012506
InChI Key NIDZUMSLERGAON-UHFFFAOYSA-N
Synonym sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
PubChem CID 171173
SMILES [H+].[Cl-].CCOC(=O)CNC

Specifications

Melting Point 122°C to 126°C
Linear Formula CH3NHCH2CO2CH2CH3·HCl
Quantity 500 g
Beilstein 3911182
Sensitivity Hygroscopic
Solubility Information Soluble in water 0.1g/mL.
IUPAC Name ethyl 2-(methylamino)acetate;hydrochloride
Formula Weight 153.61
Percent Purity ≥98%
Chemical Name or Material Sarcosine ethyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.