missing translation for 'onlineSavingsMsg'
Learn More

SAR407899, MedChemExpress

Product Code. p-200100761
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16476876 100 mg 100mg
16449297 5 mg 5mg
16426748 10 mg 10mg
16422827 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16476876

Brand: Medchem Express HY15687A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR407899 is a selective, potent and ATP-competitive ROCK inhibitor, with an IC50 of 135 nM for ROCK-2, and Kis of 36 nM and 41 nM for human and rat ROCK-2, respectively.

SAR407899 is a selective, potent and ATP-competitive ROCK inhibitor, with an IC50 of 135 nM for ROCK-2, and Kis of 36 nM and 41 nM for human and rat ROCK-2, respectively.

Specifications

Chemical Name or Material SAR407899
CAS 923359-38-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H16N2O2
Quantity 100 mg
Solubility Information DMSO : 6 mg/mL (24.56 mM; Need warming)
SMILES O=C(NC=C1)C(C1=C2)=CC=C2OC3CCNCC3
Molecular Weight (g/mol) 244.29
Formula Weight 244.29
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.