missing translation for 'onlineSavingsMsg'
Learn More

SAR-20347, MedChemExpress

Product Code. p-200097585
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16452676 25 mg 25mg
16444486 100 mg 100mg
16435947 5 mg 5mg
16413218 10 mg 10mg
16409766 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16452676

Brand: Medchem Express HY100895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR-20347 is an inhibitor of TYK2, JAK1, JAK2 and JAK3 with IC50s of 0.6, 23, 26 and 41 nM, respectively.

SAR-20347 is an inhibitor of TYK2, JAK1, JAK2 and JAK3 with IC50s of 0.6, 23, 26 and 41 nM, respectively.

Specifications

Chemical Name or Material SAR-20347
CAS 1450881-55-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H18ClFN4O4
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (224.80 mM; Need ultrasonic and warming)
SMILES O=C(C1=C(NC2=CC=C(C(N3CCOCC3)=O)C=C2)OC(C4=C(F)C=CC=C4Cl)=N1)N
Molecular Weight (g/mol) 444.84
Formula Weight 444.84
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.