missing translation for 'onlineSavingsMsg'
Learn More

SAR-20347, MedChemExpress

Product Code. 16435947
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435947

Brand: Medchem Express HY100895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR-20347 is an inhibitor of TYK2, JAK1, JAK2 and JAK3 with IC50s of 0.6, 23, 26 and 41 nM, respectively.

SAR-20347 is an inhibitor of TYK2, JAK1, JAK2 and JAK3 with IC50s of 0.6, 23, 26 and 41 nM, respectively.

Specifications

Chemical Name or Material SAR-20347
CAS 1450881-55-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H18ClFN4O4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (224.80 mM; Need ultrasonic and warming)
SMILES O=C(C1=C(NC2=CC=C(C(N3CCOCC3)=O)C=C2)OC(C4=C(F)C=CC=C4Cl)=N1)N
Molecular Weight (g/mol) 444.84
Formula Weight 444.84
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.