Learn More
Sapropterin dihydrochloride, MedChemExpress
Product Code. 16478298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478298 10 mg 10mg
16454187 50 mg 50mg
16438036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16478298 Supplier Medchem Express Supplier No. HYA0124A
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
MedChemExpress Sapropterin ((6R)-BH4) dihydrochloride is a synthetic form of BH4 that is approved for the treatment of BH4 responsive PKU.
Sapropterin ((6R)-BH4) dihydrochloride is a synthetic form of BH4 that is approved for the treatment of BH4 responsive PKU.
Specifications
Chemical Name or Material Sapropterin dihydrochloride
Physical Form Solid
CAS 69056-38-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C9H17Cl2N5O3
For Use With (Application) COVID-19-immunoregulation
Synonym (6R)-BH4 dihydrochloride(6R)-Tetrahydro-L-biopterin dihydrochloride
Solubility Information H2O : 100 mg/mL (318.30 mM; Need ultrasonic)
SMILES O=C1N=C(N)NC2=C1N[C@@H]([C@@H](O)[C@@H](O)C)CN2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 314.17
Formula Weight 314.17
Percent Purity 98.0%
Grade Research