missing translation for 'onlineSavingsMsg'
Learn More

SAPCD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15937524
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15937524 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15937524 Supplier Invitrogen Supplier No. PA549539

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SAPCD2 Polyclonal Antibody

SAPCD2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPCD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SAPCD2
Gene Accession No. Q86UD0
Gene Alias 2010317E24Rik; 6030458L21Rik; AL033337; ang; C9orf140; p42.3; Protein Ang; protein C9orf140; protein C9orf140 homolog; SAPCD2; suppressor APC domain containing 2; suppressor APC domain-containing protein 2; TS/MDEP; tumor specificity and mitosis phase-dependent expression protein
Gene Symbols SAPCD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 333-367 amino acids from human SAPCD2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89958
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.