Learn More
Notrax Sanitop™ Rubber Duckboard with Moulded Edges
Product Code. 17702218
Click to view available options
Length (Metric):
152 cm
297 cm
594 cm
Unit Size:
Each
Product Code. 17702218
Brand: Notrax 562S0320BL
Only null left
Add to basket
Add to basket
Rubber duckboards provide a comfortable and safe working platform in wet areas with its aggressive slip resistant surface. Large holes and raised studs allow fluids in wet areas or debris in dry areas to fall through leaving the surface clear.
  • Provides basic comfort and traction in wet areas with large drainage holes and raised studs for aeration and to allow fluids and debris to fall through, leaving the surface clear
  • Moulded bevelled edges on all 4 sides to prevent tripping and allow cart access
  • Rubber duckboards provide a comfortable and safe working platform in wet areas with its aggressive slip-resistant surface
  • Large holes and raised studs allow fluids in wet areas or debris in dry areas to fall through leaving the surface clear
  • Cleaning with detergent and water-jet is recommended for extended service life
Specifications
Color Black
Length (English) 19.5 ft.
Length (Metric) 594 cm
Material Natural Rubber
Type Industrial and Commercial Matting
Width (Metric) 91 cm