Learn More
Notrax Sani-Master™ Disinfecting Foot Bath with Drying Entrance Mat, Charcoal/Black
Product Code. 17722028
Change view
Click to view available options
Length (Metric):
105 cm
153 cm
200 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
17722028 200 cm Each
17702028 105 cm Each
17712028 153 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17722028 Supplier Notrax Supplier No. 348S0037CH
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Notrax Sani-Master™ 19-mm thick extendable mat cleans, sanitizes and dries shoes, recommended at entrances to prevent contamination. As traffic passes, rubber tips bend to immerse soles in disinfectant solution, absorbing mat section cleans and dries shoes to prevent slips.
  • Combining three of Notrax™ matting features of sanitizing, absorbing and modularity into a complete disinfection entrance mat solution, Sani-Master™ is a 19 mm thick extendable mat that cleans, sanitizes and dries shoes
  • Traditionally used in the food industry, Sani-Master™ has become mainstream as part of general hygiene protocols and is recommended at entrances of schools, hotels, gyms, and offices, as well as pharmacies, health care facilities, and hospitals to prevent contamination from entering the building
  • Section 1 incorporates the Sani-Trax™ disinfection foot bath with flexible bristles that bend to immerse soles in disinfectant solution to wash and disinfect shoes as traffic passes
  • Section 2 is an extendable drying zone to dry shoes and prevent slips with patented connection system that is seamless to avoid changes in height or disconnection of the mats
  • Section 3 is an end drying zone with integrated bevelled edges to prevent tripping
  • The drying carpets can be fully customized with a logo, design or used for communication of hygiene protocols such as hand disinfection and social distancing
Specifications
Color Charcoal/Black
Length (English) 6.6 ft.
Length (Metric) 200 cm
Material Natural Rubber
Type Industrial and Commercial Matting
Width (Metric) 91 cm