Learn More
Vitlab™ SAN Multi-purpose container
Product Code. 10393091
Change view
Click to view available options
Capacity (Metric):
4000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10393091 4000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10393091 Supplier Vitlab™ Supplier No. 36491
  • ONLINE EXCLUSIVE PRICE
    362.00 NOK
    288.09 NOK / Pack of 1
    Save 73.91 NOK
    20% Off
Estimated Shipment: 16-12-2026
Includes fitted lid. Vitlab™ SAN Multi-purpose container is ideal for dust-proof storage of small components, instruments and utensils.
  • Crystal clear color
  • With fitted lid 
  • Planar bottom inside
  • Reinforced outside edge for stable placement
  • Ideal for dust-proof storage of small components, instruments and utensils
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 4000 mL
Closure Type Lid
Color Clear
Shape Rectangular
For Use With (Application) Dust-Proof Storage of Small Components, Instruments and Utensils
Product Type Storage Container
Quantity 1 Each
Style Reinforced Edges
Dimensions (L x W x H) 340 x 230 x 94 mm
Length (Metric) 340 mm
Width (Metric) 230 mm
Height (Metric) 94 mm