Learn More
SAMSN1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13063-1-AP-20UL
 View more versions of this product
Product Code. 16861094
  • 1710.33 NOK / 20µL
Description
The SAMSN1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SAMSN1. This antibody recognizes human, rat antigen. The SAMSN1 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
SAMSN1
Polyclonal
Unconjugated
20 μL
64092
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
827 μg/mL
Rabbit
Primary
Human, Rat