Learn More
SAMSN1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13063-1-AP-20UL
Description
The SAMSN1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SAMSN1. This antibody recognizes human, rat antigen. The SAMSN1 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| SAMSN1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 64092 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 827 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat |