Learn More
Sampling manifold accessory for HyPerforma™ Single-Use Bioreactors, Single pack - includes Luer Locks
Product Code. 17908201 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 manifold/single-pack
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17908201 1 manifold/single-pack Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17908201 Supplier Thermo Scientific Supplier No. SH30845.01
  • ONLINE EXCLUSIVE PRICE
    7175.00 NOK
    5955.50 NOK / Pack of 5
    Save 1219.50 NOK
    16% Off
 Restricted Item
In Stock
The Sterile Sampling Manifold with Luer Locks (single-pack) is an accessory for the Thermo Scientific™ HyPerforma™ Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
The Sterile Sampling Manifold with Luer Locks (single-pack) is an accessory for the Thermo Scientific™ HyPerforma™ Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 manifold/single-pack
Product Line HyPerforma
For Use With (Equipment) Single-Use Bioreactor