Learn More
Thermo Scientific™ Sampling Manifold Accessory for HyPerforma™ Single-Use Bioreactors, with Luer locks
Product Code. 17889756 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
50/100 mL
Quantity:
1 Ea.
10/Pk.
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity unitSize
17889756 50/100 mL 1 Ea. Pack of 10
17809766 50/100 mL 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17889756 Supplier Thermo Scientific™ Supplier No. SH31263.01
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
The Sterile Sampling Manifold is an accessory for the HyPerforma Single-Use Bioreactors and is available to weld onto the sample line to take sample sets.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50/100 mL
Quantity 1 Ea.
Tubing Style C-Flex