Learn More
Dräger™ Sampling Hoses and Tools for Dräger™ accuro™ Bellows Pumps
Specifications
Specifications
| For Use With (Equipment) | Accuro Bellows Pump |
| Type | Extension Hose |
| Includes | Adapter |
| Length (English) | 33 ft. |
| Length (Metric) | 10 m |