Learn More
BOLA Sampling Bags
Product Code. 10641132
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
5 L
10 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10641132 5 L Each
10768622 1 L Each
10680552 2 L Each
10745982 10 L Each
4 options
Product Code. 10641132
Brand: BOLA A22370
Only null left
Add to basket
Add to basket
Sampling Bags PVF, 2 connectors
  • Transparent
  • Non-porous
  • Without plasticisers
  • With 2 exchangeable ports made of PTFE.
Specifications
Capacity (Metric) 5 L
Color White
Length (Metric) 250 mm
Width (Metric) 350 mm