Learn More
Cole-Parmer™ Kynar™ Dual-Valve PVDF Gas Sampling Bags
Product Code. 11750099 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
0.3 L
1.6 L
3.8 L
37.7 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11750099 1.6 L Each
11715868 0.3 L Each
12510708 3.8 L Each
11750009 37.7 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11750099 Supplier Cole-Parmer™ Supplier No. 0141051
  • ONLINE EXCLUSIVE PRICE
    772.00 NOK
    739.18 NOK / Pack of 1
    Save 32.82 NOK
    4% Off
View all promotions
Estimated Shipment: 13-01-2027
Use these bags made from corrosive-resistant Kynar™ for all your application needs. Standard Kynar™ PVDF Gas Sampling Bags feature strong seams for added durability.
Use these bags made from corrosive-resistant KynarTM for all your application needs. Standard KynarTM PVDF Gas Sampling Bags feature strong seams for added durability.
  • Double-wide seamed bags have both on/off and septum valves for convenient mixing
  • Max temp: 160°F (71°C)
  • Individually sealed in a dust-protected outer bag that also blocks out sunlight
  • Single-wide seam
  • Available in 2-mil and 4-mil thick style
Specifications
Material PVDF
Temperature Range (Metric) 71°C
Thickness (English) 3 mil
Dimensions (L x W) 228.6 x 228.6mm
Product Type Gas Sampling Bag
Capacity (Metric) 1.6 L
Length (Metric) 228.6 mm
Width (Metric) 228.6 mm