Learn More
Lonza™ Sample Vials, Pyrogen Free
Product Code. 11640131
Change view
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11640131 10 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11640131 Supplier Lonza™ Supplier No. 80507U
Estimated Shipment: 14-12-2026
For transportation or storage of samples for endotoxin analysis. Lonza™ Sample Vials, Pyrogen Free are 10mL glass endotoxin-free sample containers with screw caps. 100/box
Sample Containers are intended for transporting product samples for endotoxin analysis or sample storage. Proper container and storage conditions need to be validated for each individual sample.
  • 10mL depyrogenated glass sample containers with screw caps
  • Certified to contain less than 0.005 EU/mL
  • To be used in applications requiring the endotoxin test
  • 100 vials/box (cardboard box with cardboard insert separators)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100/Box
Capacity (Metric) 10 mL
Color Clear, with black cap
Product Type Sample Vials with Lids
Material Depyrogenated Glass
For Use With (Application) Endotoxin Analysis