Description
Polycarbonate cryovial with screw-on cap, 160mm x 121mm, 15mL. Holds grinding media up to 3/8in and fits SPEX SamplePrep homogeniser 2197 holder and 2661 cryo-block. For use with 2199 large clamp assembly. Grinding load per vial 6mL. Sold as a pack of 30 vials.
- Holds grinding load of 6mL per vial
- Polycarbonate so suitable for cryogenic grinding
Specifications
Specifications
| Diameter (Metric) Outer | 15.8 mm |
| Material | Polycarbonate |
| Volume (Metric) | 15 mL |
| Includes | With Screw on Cap |
| No. per Pack | 30 |
| Closure Type | Screw Cap |
| Quantity | 30 Pk. |
| For Use With (Equipment) | 2199 Large Clamp Assembly |
| Height (Metric) | 101.6 mm |