Learn More
Sample loop (7125, 7010) SST 1/16 inch O.D.,0.030 inch (0.76mm) I.D. 500µL
Brand:  N/A 02016-39
Sample loop (7125, 7010) SST 1/16 inch O.D.,0.030 inch (0.76mm) I.D. 500µL
Product Code. 13070193
  • ONLINE EXCLUSIVE PRICE
    1430.00 NOK
    1245.42 NOK / Each
    Save 184.58 NOK
    12% Off