Learn More
BRAND™ Sample Cups, PS, clear, for Technicon-Analyzer
Product Code. p-10611132 Shop All Brand Products
Click to view available options
Volume (Metric):
1.5 mL
2.0 mL
4 mL
Unit Size:
Pack of 1000
Product Code. 10165261
Brand: BRAND™ 115015
Only null left
Add to basket
Add to basket
Transparent polystyrene. For use with Technicon Analyzer.
  • Sample vial
  • Polystyrene construction
  • Glass clear
  • IVD 1.5 mL
  • For use with Technicon Analyzer
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Diameter (Metric) Outer 12.2 mm
Material Polystyrene
Volume (Metric) 1.5 mL
No. per Pack 1000
Closure Material Polystyrene
Height (Metric) 22.7 mm
Quantity 1000 Pk.
For Use With (Equipment) Technicon Analyzer