Learn More
WTW™ Sample Bottle
Product Code. 10528591
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
510 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10528591 1000 mL Each
10518591 500 mL Each
10364831 510 mL Each
10283291 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10528591 Supplier WTW™ Supplier No. 209104
  • ONLINE EXCLUSIVE PRICE
    522.00 NOK
    515.46 NOK / Pack of 1
    Save 6.54 NOK
    1% Off
View all promotions
Estimated Shipment: 01-12-2026
Replacement sample bottles and other glassware as well as useful accessories for the expansion of application of measuring systems for different parameters.WTW offers a large assortment of glass ware and other products to replace, supplement and to extend your applications.
TRUSTED_SUSTAINABILITY
Specifications
Description Sample bottle
Volume (Metric) 1000 mL