missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD9L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25173-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein has not been specifically defined.Specifications
SAMD9L | |
Polyclonal | |
Unconjugated | |
SAMD9L | |
C7orf6, DRIF2, KIAA2005, SAMD9L, UEF, UEF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
219285 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IVG5 | |
SAMD9L | |
SAMD9L Fusion Protein Ag18541 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |