Description
The function of this protein remains unknown.
Specifications
Specifications
| Antigen | SAMD3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1A11F2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SAMD3 |
| Gene Accession No. | Q8N6K7 |
| Gene Alias | SAMD3 |
| Show More |