Learn More
Thermo Scientific™ Samco™ Extra Long Transfer Pipettes
Description
Perfect for research applications, these Extra-Long Transfer Pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, and autoclaving and prevents cross-contamination. They allow safe and easy sampling from large containers.
- Excellent for research applications
- General-purpose pipettes are ideal for routine sampling of liquids
- Well suited for use with large containers
Specifications
Specifications
| Bulb Draw (Metric) | 2.3 mL |
| Diameter (Metric) Outer | 5.0 mm |
| Material | LDPE |
| Sterility | Sterile |
| Drops/mL | 25 |
| Graduated | No |
| Length (English) | 9 in. |
| Quantity | 400/Pk 8Pk/Cs 3200/Cs |
| Packaging Type | Individually Wrapped |
| Volume (Metric) | 5.6 mL |
| Show More |